Order Entry
Italy
ContactUsLinkComponent
Methyl-4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate
Methyl-4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate
Catalog # APOSOR29670-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl-4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate
Catalog # APOSOR29670-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :247.0891 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:175276-98-9
Formula:C₈H₁₁BrN₂O₂
MDL Number:MFCD00173775