Order Entry
Italy
ContactUsLinkComponent
Di-tert-butyl pyrocarbonate 99%
Flammable
Corrosive
Toxic
Di-tert-butyl pyrocarbonate 99%
Catalog # APOSOR28895-250G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Corrosive
Toxic
Di-tert-butyl pyrocarbonate 99%
Catalog # APOSOR28895-250G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :218.25 g/mol
Boiling Pt:56…57 °C/0.5mmHg
Melting Pt:23 °C
Density:0.95 g/cm³ (25 °C)
Flash Pt:37 °C
Storage Temperature:Refrigerator
EINECS:246-240-1
UN:2930
ADR:6.1+4.1,II
 :GHS02
 :GHS05
 :GHS06
CAS Number:24424-99-5
Formula:C₁₀H₁₈O₅
MDL Number:MFCD00008805