
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
Catalog # APOSOR28603-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:92…95 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
CAS Number:4462-55-9
Formula:C₁₁H₆Cl₃NO₂
MDL Number:MFCD00052556