
1,8-Naphthalenedicarboxylic anhydride 98%
Catalog # APOSOR28586-250G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 g
- Seq:3545
Specifications
About this item
:
Melting Pt:267…273 °C
Flash Pt:272 °C
:198.18 g/mol
EINECS:201-380-2
:GHS07
Storage Temperature:Ambient
CAS Number:81-84-5
Formula:C₁₂H₆O₃
MDL Number:MFCD00006925