Order Entry
Italy
Orders LinkContactUsLinkComponent
N-Hydroxysuccinimide 99%

N-Hydroxysuccinimide 99%

Irritant
Supplier:   Apollo Scientific
N-Hydroxysuccinimide 99%
Supplier:  Apollo Scientific
Certificates
N-Hydroxysuccinimide 99%

3 Options Available Below

Product Details & Documents
Synonyms:  1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
  • CAS Number:6066-82-6
  • Formula:C₄H₅NO₃
  • MDL Number:MFCD00005516
  • Molecular Weight:115.09 g/mol
  • Melting Pt:94…99 °C
  • Storage Temperature:Ambient
  • EINECS:228-001-3

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.