Order Entry
Italy
ContactUsLinkComponent
 
Irritant
1,1-Dioxidotetrahydrothien-3-ylamine hydrochloride
Catalog # APOSOR28153-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,1-Dioxidotetrahydrothien-3-ylamine hydrochloride
Catalog # APOSOR28153-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :171.65 g/mol
Melting Pt:208…209 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:51642-03-6
Formula:C₄H₁₀ClNO₂S
MDL Number:MFCD00456584