
4-(Morpholin-4-yl)-3-nitroaniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5367-65-7
:
Formula:C₁₀H₁₃N₃O₃
MDL Number:MFCD05257026
:223.23 g/mol
Melting Pt:131…133 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together