Order Entry
Italy
ContactUsLinkComponent
Ethyl-3-aminopropanoate hydrochloride
Irritant
Ethyl-3-aminopropanoate hydrochloride
Catalog # APOSOR26826-10G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-3-aminopropanoate hydrochloride
Catalog # APOSOR26826-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :153.6 g/mol
Melting Pt:67…72 °C
EINECS:224-203-0
 :GHS07
Storage Temperature:Ambient
CAS Number:4244-84-2
Formula:C₅H₁₁NO₂·HCl
MDL Number:MFCD00012909