
2-Chloro-4-iodopyridine-3-carboxaldehyde
Catalog # APOSOR2658-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:267.45 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:153034-90-3
Formula:C₆H₃ClINO
MDL Number:MFCD03095294