
Diethyl cyclopropane-1,1-dicarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1559-02-0
:
Formula:C₉H₁₄O₄
MDL Number:MFCD00001283
:186.21 g/mol
Boiling Pt:94…96 °C
Density:1.055 g/cm³ (25 °C)
Flash Pt:87 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together