Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dimethoxy-3-pyridineboronic acid
2,6-Dimethoxy-3-pyridineboronic acid
2,6-Dimethoxy-3-pyridineboronic acid
APOSOR2616-250MG
:
Apollo Scientific
:
2,6-Dimethoxy-3-pyridineboronic acid
APOSOR2616-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
182.97 g/mol
:
100…117 °C
:
GHS07
:
Refrigerator
:
221006-70-8
:
C₇H₁₀BNO₄
:
MFCD03788239