
2-Chloro-1-(5-chloro-3-methyl-1-benzo[b]thien-2-yl)ethan-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:156801-47-7
:
Formula:C₁₁H₈Cl₂OS
MDL Number:MFCD00052305
:259.16 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together