
4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid
Catalog # APOSOR25990-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.0360 g/mol
Melting Pt:230…232 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5775-88-2
Formula:C₆H₇BrN₂O₂
MDL Number:MFCD02090873