Order Entry
Italy
ContactUsLinkComponent
(5-Carboxypentyl)triphenylphosphonium bromide 97%
(5-Carboxypentyl)triphenylphosphonium bromide 97%
Catalog # APOSOR25662-100G
Supplier:  Apollo Scientific
CAS Number:  
(5-Carboxypentyl)triphenylphosphonium bromide 97%
Catalog # APOSOR25662-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :457.36 g/mol
Storage Temperature:Ambient
CAS Number:50889-29-7
Formula:C₂₄H₂₆BrO₂P
MDL Number:MFCD00055556