
2-(Methylthio)benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3724-10-5
:
Formula:C₈H₈O₂S
MDL Number:MFCD00029934
:168.22 g/mol
Melting Pt:169…173 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together