
4,7-Dimethylpyrazolo[5,1-c]-[1,2,4]triazine-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175137-58-3
:
Formula:C₈H₈N₄O₂
MDL Number:MFCD00067873
:192.18 g/mol
Melting Pt:166…168 °C
Storage Temperature:Ambiente
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together