

Morpholine-4-carboximidamide hydroiodide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:102392-87-0
:
Formula:C₅H₁₂IN₃O
MDL Number:MFCD00114180
:257.08 g/mol
Melting Pt:143…144 °C
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Frequently Bought Together