
1-Methylcyclopropanecarboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6914-76-7
:
Formula:C₅H₈O₂
MDL Number:MFCD00001290
:100.12 g/mol
Boiling Pt:183…185 °C
Melting Pt:30 °C
Flash Pt:84 °C
Storage Temperature:Ambient
EINECS:230-020-7
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together