Methyl-4-bromo-3-hydroxythiophene-2-carboxylate
Catalog # APOSOR24351-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:237.08 g/mol
Storage Temperature:Ambient
CAS Number:95201-93-7
Formula:C₆H₅BrO₃S
MDL Number:MFCD00052081