Order Entry
Italy
ContactUsLinkComponent
Methyl-4-aminothiophene-3-carboxylate hydrochloride 96%
Irritant
Methyl-4-aminothiophene-3-carboxylate hydrochloride 96%
Codice catalogo APOSOR24349-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-4-aminothiophene-3-carboxylate hydrochloride 96%
Codice catalogo APOSOR24349-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.65 g/mol
Melting Pt:183…185 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:39978-14-8
Formula:C₆H₈ClNO₂S
MDL Number:MFCD00068149