
1,8-Octanediol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:629-41-4
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00002989
:146.23 g/mol
Boiling Pt:71…73 °C/20mmHg
Melting Pt:59…61 °C
Flash Pt:148 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together