



2-Mercaptoacetamide 10% (w/v) in methanolic ammonia
Catalog # APOSOR24255-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:91.13 g/mol
Boiling Pt:64 °C
Density:0.89 g/cm³ (25 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
UN:3286
ADR:3+6.1+8,II
:GHS02
:GHS05
:GHS06
:GHS08
CAS Number:758-08-7
Formula:C₂H₅NOS
MDL Number:MFCD00068159