
4-Acetyl-4'-methoxydiphenyl ether
Catalog # APOSOR24033-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.28 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:54916-28-8
Formula:C₁₅H₁₄O₃
MDL Number:MFCD00052928