
2-Phenyl-1,3-oxazole-4-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23012-16-0
:
Formula:C₁₀H₇NO₃
MDL Number:MFCD03236685
Melting Pt:211…212 °C
:189.17 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together