Order Entry
Italy
ContactUsLinkComponent
6-Phenoxy-3-formylpyridine
Irritant
6-Phenoxy-3-formylpyridine
Catalog # APOSOR23319-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Phenoxy-3-formylpyridine
Catalog # APOSOR23319-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :199.21 g/mol
Melting Pt:102…103 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:173282-69-4
Formula:C₁₂H₉NO₂
MDL Number:MFCD02681950