Order Entry
Italy
ContactUsLinkComponent
4-(1H-Pyrazol-1-yl)benzaldehyde 97%
Irritant
4-(1H-Pyrazol-1-yl)benzaldehyde 97%
Catalog # APOSOR23291-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(1H-Pyrazol-1-yl)benzaldehyde 97%
Catalog # APOSOR23291-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :172.1833 g/mol
Melting Pt:82…83 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:99662-34-7
Formula:C₁₀H₈N₂O
MDL Number:MFCD02681938