Order Entry
Italy
ContactUsLinkComponent
tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate 95+%
tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate 95+%
Catalog # APOSOR2326-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate 95+%
Catalog # APOSOR2326-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.36 g/mol
Storage Temperature:Ambient
CAS Number:1031927-12-4
Formula:C₁₄H₂₄N₂O₃
MDL Number:MFCD09998742