Order Entry
Italy
ContactUsLinkComponent
2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester 95+%
Irritant
2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester 95+%
Catalog # APOSOR2324-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester 95+%
Catalog # APOSOR2324-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :266.34 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1031927-11-3
Formula:C₁₄H₂₂N₂O₃
MDL Number:MFCD09998740