Order Entry
Italy
ContactUsLinkComponent
1,4-Diazepan-5-one hydrochloride 98%
Irritant
1,4-Diazepan-5-one hydrochloride 98%
Catalog # APOSOR2299-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,4-Diazepan-5-one hydrochloride 98%
Catalog # APOSOR2299-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :150.6066 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:208245-76-5
Formula:C₅H₁₁ClN₂O
MDL Number:MFCD09864983