Order Entry
Italy
ContactUsLinkComponent
 
Irritant
1-(1-Adamantyl)ethylamine hydrochloride
Catalog # APOSOR22496-10G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(1-Adamantyl)ethylamine hydrochloride
Catalog # APOSOR22496-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :215.77 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1501-84-4
Formula:C₁₂H₂₁N
MDL Number:MFCD00072023