Inserisci ordine
Italy
ContactUsLinkComponent
2,4-Dibromobenzoic acid 97%
Irritant
2,4-Dibromobenzoic acid 97%
Catalog # APOSOR2171-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dibromobenzoic acid 97%
Catalog # APOSOR2171-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.92 g/mol
Melting Pt:50…52 °C
 :GHS07
Storage Temperature:Ambiente
CAS Number:611-00-7
Formula:C₇H₄Br₂O₂
MDL Number:MFCD00234253