3-Chlorobenzo[b]thiophene-2-carboxamide
Catalog # APOSOR21581-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.67 g/mol
Storage Temperature:Ambient
CAS Number:21211-09-6
Formula:C₉H₆ClNOS
MDL Number:MFCD00067793