Order Entry
Italy
ContactUsLinkComponent
2,6-Dibromo-4-cyclohexylaniline
2,6-Dibromo-4-cyclohexylaniline
Catalog # APOSOR21303-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dibromo-4-cyclohexylaniline
Catalog # APOSOR21303-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :333.07 g/mol
Storage Temperature:Ambient
CAS Number:175135-11-2
Formula:C₁₂H₁₅Br₂N