
(1H-Imidazol-4-yl)methanol hydrochloride 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:32673-41-9
:
Formula:C₄H₇ClN₂O
MDL Number:MFCD00012697
:134.57 g/mol
Melting Pt:108…112 °C
EINECS:251-150-0
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together