Order Entry
Italy
ContactUsLinkComponent
4-(2,4-Dinitrophenyl)morpholine
4-(2,4-Dinitrophenyl)morpholine
Catalog # APOSOR200048-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(2,4-Dinitrophenyl)morpholine
Catalog # APOSOR200048-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :253.22 g/mol
Melting Pt:117…118 °C
Storage Temperature:Ambient
CAS Number:39242-76-7
Formula:C₁₀H₁₁N₃O₅
MDL Number:MFCD00193779