Ethyl-1-(2-chloropyrimidin-4-yl)piperidine-3-carboxylate
Catalog # APOSOR200046-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.73 g/mol
Storage Temperature:Ambient
CAS Number:1347757-99-6
Formula:C₁₂H₁₆ClN₃O₂
MDL Number:MFCD22126068