Order Entry
Italy
ContactUsLinkComponent
4-(2,4-Dinitrophenoxy)benzaldehyde
4-(2,4-Dinitrophenoxy)benzaldehyde
Catalog # APOSOR200041-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(2,4-Dinitrophenoxy)benzaldehyde
Catalog # APOSOR200041-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :288.22 g/mol
Storage Temperature:Ambient
CAS Number:794-65-0
Formula:C₁₃H₈N₂O₆
MDL Number:MFCD00579130