Di-tert-butyl-2-chloropyridin-3-ylimidodicarbonate
Catalog # APOSOR200040-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:328.8 g/mol
Storage Temperature:Ambient
CAS Number:749265-07-4
Formula:C₁₅H₂₁ClN₂O₄
MDL Number:MFCD21496441