
3-Chloro-4-formylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:72990-37-5
:
Formula:C₆H₄ClNO
MDL Number:MFCD06200712
:141.56 g/mol
Melting Pt:58…62 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together