
4-Chlorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74-11-3
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00002531
:156.57 g/mol
Boiling Pt:275 °C
Melting Pt:238…241 °C
Density:1.54 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:200-805-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together