Order Entry
Italy
ContactUsLinkComponent
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 97%
Irritant
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 97%
Catalog # APOSOR18601-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 97%
Catalog # APOSOR18601-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.622 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:188781-08-0
Formula:C₈H₉ClN₂O₂
MDL Number:MFCD03425963