1-Hydroxy-2-naphthaldehyde
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:574-96-9
- Formula:C₁₁H₈O₂
- MDL Number:MFCD00092325
- Molecular Weight:172.1800 g/mol
- Melting Pt:56 °C
- Storage Temperature:Ambient
6 Options Available Below
Product Details & Documents
- CAS Number:574-96-9
- Formula:C₁₁H₈O₂
- MDL Number:MFCD00092325
- Molecular Weight:172.1800 g/mol
- Melting Pt:56 °C
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



