1-Methylpiperazine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:109-01-3
:
Formula:C₅H₁₂N₂
MDL Number:MFCD00005966
:100.16 g/mol
Boiling Pt:137…139 °C
Melting Pt:–5 °C
Density:0.903 g/cm³ (25 °C)
Flash Pt:39 °C
Storage Temperature:Ambient
EINECS:203-639-5
UN:2734
ADR:8+3,II
:GHS02
:GHS05
:GHS06
Frequently Bought Together