Order Entry
Italy
ContactUsLinkComponent
3-[1,1'-Biphenyl]-4-yl-1H-pyrazole
3-[1,1'-Biphenyl]-4-yl-1H-pyrazole
Catalog # APOSOR183455-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[1,1'-Biphenyl]-4-yl-1H-pyrazole
Catalog # APOSOR183455-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.28 g/mol
Storage Temperature:Ambient
CAS Number:446276-22-8
Formula:C₁₅H₁₂N₂
MDL Number:MFCD02755978