
Disodium-3-hydroxy-2,7-naphthalenedisulphonate, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:135-51-3
:
Formula:C₁₀H₆O₇S₂.2Na
MDL Number:MFCD00065915
:348.25 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together