
1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:454482-11-2
:
Formula:C₁₂H₂₂BNO₂
MDL Number:MFCD11506069
:223.12 g/mol
Melting Pt:72…76 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together