
(R)-3-Methylmorpholine hydrochloride 97%
Codice catalogo APOSOR17129-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.6100 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:953780-78-4
Formula:C₅H₁₂ClNO
MDL Number:MFCD18382515