
Ethyl-5-nitro-1H-pyrrole-2-carboxylate
Catalog # APOSOR17066-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:184.15 g/mol
Melting Pt:146 °C
:GHS07
Storage Temperature:Ambient
CAS Number:36131-46-1
Formula:C₇H₈N₂O₄
MDL Number:MFCD09025771