
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3680-71-5
:MFCD01686814
Formula:C₆H₅N₃O
MDL Number:MFCD08234923
:135.13 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together