
1-Methoxy-1,4-cyclohexadiene, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2886-59-1
:
Formula:C₇H₁₀O
MDL Number:MFCD00001536
Boiling Pt:148…150 °C
Density:0.94 g/cm³ (25 °C)
Flash Pt:36 °C
Storage Temperature:Ambient
EINECS:220-747-8
UN:1993
ADR:3,III
:GHS02
Frequently Bought Together